Skip to Content
Merck
All Photos(2)

Documents

GF65888274

Nickel

foil, 0.5m coil, thickness 0.30mm, as rolled, 99.98%

Synonym(s):

Nickel, NI000569

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99.98%

form

foil

manufacturer/tradename

Goodfellow 658-882-74

resistivity

6.97 μΩ-cm, 20°C

diam. × thickness

0.5 m × 0.30 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Schulz et al.
Physics in medicine and biology, 35(11), 1563-1574 (1990-11-01)
The thermal defects of A-150 plastic and graphite referenced to aluminium were determined for 800 keV protons scattered by a 2 micron nickel foil (mean transmitted energy = 550 keV). Composite cores of Al/A-150, graphite/A-150 and Al/graphite which could be
Kwanghyun Yoo et al.
Nanotechnology, 24(20), 205302-205302 (2013-04-20)
We firstly introduce a facile method for the site-specific direct physical exfoliation of few-layer graphene sheets from cheap and easily enlargeable graphite grown on a Ni foil using an optimized polydimethylsiloxane (PDMS) stamp. By decreasing the PDMS cross-linking time, the
G E van Dorssen et al.
Journal of X-ray science and technology, 3(2), 109-117 (1992-01-01)
The structure of Ni-C multilayer and single nickel layer samples has been analyzed before and after annealing, using two techniques: fluorescence EXAFS (F1EXAFS) at the Ni-K. edge and CuKα reflection. Annealing at a temperature of 450°C resulted in a change
F E Cecil et al.
The Review of scientific instruments, 81(10), 10D326-10D326 (2010-11-09)
We have examined the observed currents in the front foils of the JET Faraday cup lost alpha particle diagnostic KA-2. In particular, we have sought to understand the currents during Ohmic plasmas for which the ion flux at the detectors
W K Winnett et al.
Talanta, 41(10), 1627-1630 (1994-10-01)
A modification of the sample introduction system for combustion total organic carbon analyzers is presented where sample is sprayed into the instrument. Thermally created sprays and nebulizer sprays are discussed. It is shown that spray introduction gives more symmetric signals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service