Skip to Content
Merck
All Photos(1)

Key Documents

CDS001582

Sigma-Aldrich

Gallium (III) nitrate, solution Ga 9-10% W/W

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
GaN3O9
CAS Number:
Molecular Weight:
255.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solution

reaction suitability

core: gallium
reagent type: catalyst

SMILES string

[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ga.3NO3/c;3*2-1(3)4/q+3;3*-1

InChI key

CHPZKNULDCNCBW-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Liq. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Natalie Gugala et al.
Antibiotics (Basel, Switzerland), 8(2) (2019-05-06)
In response to the occurrence of antibiotic resistance, there has been rapid developments in the field of metal-based antimicrobials. Although it is largely assumed that metals provide broad-spectrum microbial efficacy, studies have shown that this is not always the case.
Natalie Gugala et al.
Genes, 10(1) (2019-01-13)
The diagnostic and therapeutic agent gallium offers multiple clinical and commercial uses including the treatment of cancer and the localization of tumors, among others. Further, this metal has been proven to be an effective antimicrobial agent against a number of
Mingshi Zhao et al.
Nanoscale, 8(37), 16670-16676 (2016-10-08)
In this work, novel CuGaS
Marta Choinska et al.
Monatshefte fur chemie, 1-7 (2021-01-26)
A new method for determination of selected heavy metals (Cd, Pb, Cu, Zn, and Ni) in honey bee venom was developed. Heavy metals are metabolized and incorporated into bee products, including honey and honey bee venom (apitoxin). Their composition reflects
Yoshihiro Tamada et al.
Journal of bioscience and bioengineering, 127(6), 710-713 (2019-01-02)
Dimethyl trisulfide (DMTS) is the main component of hineka, an off-flavor generated in sake during storage. Genshu, or undiluted sake, is usually diluted with water during warimizu, the process of adjusting the alcohol content of sake. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service