900384
2′-Deoxyguanosine-13C10,15N5 5′-monophosphate disodium salt solution
100 mM (in 5mM Tris HCl / H2O), ≥98 atom %, ≥95% (CP)
Synonym(s):
2′-Deoxy-5′-guanylic-13C10,15N5 acid disodium salt solution, dGMP-13C10,15N5 disodium salt solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
13C10H1215N5Na2O7P
Molecular Weight:
406.08
UNSPSC Code:
41106305
NACRES:
NA.12
Recommended Products
isotopic purity
≥98 atom %
Assay
≥95% (CP)
form
liquid
concentration
100 mM (in 5mM Tris HCl / H2O)
technique(s)
bio NMR: suitable
mass shift
M+15
storage temp.
−20°C
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service