Skip to Content
Merck
All Photos(1)

Key Documents

749052

Sigma-Aldrich

Chromium

sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis

Synonym(s):

Chromium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis

form

solid

reaction suitability

core: chromium

resistivity

12.7 μΩ-cm, 20°C

diam. × thickness

3.00 in. × 0.125 in.

bp

2672 °C (lit.)

mp

1857 °C (lit.)

density

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

InChI key

VYZAMTAEIAYCRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sputtering is a process whereby atoms are ejected from a solid target material due to bombardment of the target by energetic particles. It is commonly used for thin-film deposition, etching and analytical techniques.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A J Vanoirbeek et al.
Journal of toxicology and environmental health. Part A, 66(4), 393-409 (2003-01-30)
Chromium-based catalysts are used for the synthesis of polyethylene, but little is known about the hazard and biomonitoring possibilities of this type of chromium for workers who may be occupationally exposed to such compounds. Therefore, the bioavailability and toxicokinetics of
B Seifert et al.
Journal of exposure analysis and environmental epidemiology, 10(2), 103-114 (2000-05-03)
The German Environmental Survey (GerES) is a large-scale population study which has repeatedly been carried out in Germany. GerES I was conducted in 1985/1986 followed by GerES IIa in 1990/1991 (West Germany) and GerES IIb in 1991/1992 (East Germany). GerES
Feng Liu et al.
Scientific reports, 3, 1554-1554 (2013-03-28)
In conventional photostimulable storage phosphors, the optical information written by x-ray or ultraviolet irradiation is usually read out as a visible photostimulated luminescence (PSL) signal under the stimulation of a low-energy light with appropriate wavelength. Unlike the transient PSL, here
A J Hart et al.
The Journal of bone and joint surgery. American volume, 95(8), 678-685 (2013-04-19)
Determining the relationship between clinical factors and engineering analysis of retrieved hip implants can help our understanding of the mechanism of device failure. This is particularly important for metal-on-metal hip arthroplasties because the most common cause of failure is unexplained.
Carola Lidén et al.
Contact dermatitis, 54(5), 233-238 (2006-05-13)
There is a great need to accurately assess skin exposure to contact allergens. We have developed a technique for assessment of skin exposure to nickel, chromium and cobalt using acid wipe sampling by cellulose wipes with 1% nitric acid. Chemical

Articles

Dye-Sensitized and Perovskite Solar Cells: Interface Engineering by Atomic Layer Deposition

A hard disk drive (HDD) is a data storage device that stores digital information by magnetizing nanosized magnets on flat disks and retrieves data by sensing the resulting magnetic field.

Spin-based electronic (spintronic) devices offer significant improvement to the limits of conventional charge-based memory and logic devices which suffer from high power usage, leakage current, performance saturation, and device complexity.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service