Skip to Content
Merck
All Photos(1)

Key Documents

536512

Sigma-Aldrich

Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene]

Synonym(s):

MEH-PPV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C18H28O2)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Band gap: 2.3 eV

fluorescence

λex 493 nm; λem 554 nm in toluene

Orbital energy

HOMO -5.3 eV 
LUMO -3 eV 

OLED Device Performance

ITO/PEDOT:PSS/MEH-PPV/Al

  • Color: orange-red
  • Max. Luminance: 220 Cd/m2
  • Max. EQE: 0.3 %
  • Turn-On Voltage: 3.8 V

ITO/PEDOT:PSS/MEH-PPV/PBO/Al
  • Color: orange-red
  • Max. Luminance: 970 Cd/m2
  • Max. EQE: 1.9 %
  • Turn-On Voltage: 2.9 V

ITO/PEDOT:PSS/MEH-PPV/PBZT/Al
  • Color: orange-red
  • Max. Luminance: 1400 Cd/m2
  • Max. EQE: 2.5 %
  • Turn-On Voltage: 2.8 V

OPV Device Performance

ITO/MEH-PPV/BBL/Al

  • Short-circuit current density (Jsc): 1.98 mA/cm2
  • Open-circuit voltage (Voc): 0.93 V
  • Fill Factor (FF): 0.47
  • Power Conversion Efficiency (PCE): 1.1 %

ITO/MEH-PPV:PC61BM(1:4)/Ca
  • Short-circuit current density (Jsc): 2 mA/cm2
  • Open-circuit voltage (Voc): 0.8 V
  • Fill Factor (FF): 0.25
  • Power Conversion Efficiency (PCE): 1.5 %

Mw/Mn

<4

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Light-emitting conjugated polymer.
MEH-PPV is a semiconductor based conjugated polymer which has good electroluminescence and excellent optoelectronic properties. It has a high quantum yield of fluorescence and it is soluble with most of the organic solvents.

Application

Conducting polymer in solar cells and carbon nanotube OLEDs. Useful in producing bright and efficient white polymeric light emitting diodes.
Filter before application.
MEH-PPV can be used as a conducting polymer in the fabrication of the electronic devices such as:
  • field effect transistors(FET)
  • photovoltaic cells
  • light emitting diodes(LED)
  • sensors for biomedical applications
  • photodiodes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complex nanoparticle of light-emitting MEH-PPV with Au: enhanced luminescence
Kim MS, et al.
ACS Nano, 3(6), 1329-1334 (2009)
A large interconnecting network within hybrid MEH-PPV/TiO2 nanorod photovoltaic devices
Zeng T, et al.
Nanotechnology, 17(21), 5387-5387 (2006)
General observation of n-type field-effect behaviour in organic semiconductors
Chua L, et al.
Nature, 434(7030), 194-194 (2005)
a new target ligand ser-glu for PePT1-overexpressing cancer imaging
Dai T, et al.
International journal of nanomedicine, 11(6378), 203-203 (2016)
Tongcheng Dai et al.
International journal of nanomedicine, 11, 203-212 (2016-01-27)
Nanoparticles functionalized with active target ligands have been widely used for tumor-specific diagnosis and therapy. The target ligands include antibodies, peptides, proteins, small molecules, and nucleic acid aptamers. Here, we utilize dipeptide Ser-Glu (DIP) as a new ligand to functionalize

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Light-Emitting Polymers

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service