Skip to Content
Merck
All Photos(1)

Key Documents

394920

Supelco

1-(Trifluoroacetyl)imidazole

for GC derivatization, LiChropur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3F3N2O
CAS Number:
Molecular Weight:
164.09
Beilstein:
608904
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
form:
liquid
Assay:
>98%

grade

for GC derivatization

Assay

>98%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.424 (lit.)

bp

137 °C (lit.)
45-46 °C/14 mmHg (lit.)

density

1.442 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)n1ccnc1

InChI

1S/C5H3F3N2O/c6-5(7,8)4(11)10-2-1-9-3-10/h1-3H

InChI key

SINBGNJPYWNUQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(Trifluoroacetyl)imidazole is a derivatizing reagent.

Application

1-(Trifluoroacetyl)imidazole was used as a derivating agent to sequentially derivatize functional groups like amino groups, in a study demonstrating trimethylsilyl esters of aromatic and aliphatic sulfonic acids.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Eagles and M.E. Knowles
Analytical Chemistry, 43, 1697-1697 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service