Merck
HPLC Analysis of Penicillin G and Phenoxymethylpenicillinic Acid on Ascentis® C18
HPLC Analysis of Penicillin G and Phenoxymethylpenicillinic Acid on Ascentis® C18 application for HPLC
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
Colonne Ascentis® C18 pour HPLC
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase
[A] 10 mM ammonium acetate (pH 4.5 acetic acid): [B] acetonitrile, (75:25, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 220 nm
injection
10 μL
sample
100 μg/mL each in mobile phase