Merck
Transcription regulation of CDKN1A (p21/CIP1/WAF1) by TRF2 is epigenetically controlled through the REST repressor complex.
Scientific reports (2017-09-16)
Tabish Hussain, Dhurjhoti Saha, Gunjan Purohit, Anirban Kar, Anand Kishore Mukherjee, Shalu Sharma, Suman Sengupta, Parashar Dhapola, Basudeb Maji, Sreekanth Vedagopuram, Nobuko T Horikoshi, Nobuo Horikoshi, Raj K Pandita, Santanu Bhattacharya, Avinash Bajaj, Jean-François Riou, Tej K Pandita, Shantanu Chowdhury
RÉSUMÉ
We observed extra-telomeric binding of the telomere repeat binding factor TRF2 within the promoter of the cyclin-dependent kinase CDKNIA (p21/CIP1/WAF1). This result in TRF2 induced transcription repression of p21. Interestingly, p21 repression was through engagement of the REST-coREST-LSD1-repressor complex and altered histone marks at the p21 promoter in a TRF2-dependent fashion. Furthermore, mutational analysis shows p21 repression requires interaction of TRF2 with a p21 promoter G-quadruplex. Physiologically, TRF2-mediated p21 repression attenuated drug-induced activation of cellular DNA damage response by evading G2/M arrest in cancer cells. Together these reveal for the first time role of TRF2 in REST- repressor complex mediated transcription repression.