Merck
  • Suppression of heat shock protein 27 using OGX-427 induces endoplasmic reticulum stress and potentiates heat shock protein 90 inhibitors to delay castrate-resistant prostate cancer.
Suppression of heat shock protein 27 using OGX-427 induces endoplasmic reticulum stress and potentiates heat shock protein 90 inhibitors to delay castrate-resistant prostate cancer.
European urology (2014-01-15)
François Lamoureux, Christian Thomas, Min-Jean Yin, Ladan Fazli, Amina Zoubeidi, Martin E Gleave
RÉSUMÉ
Although prostate cancer responds initially to androgen ablation therapies, progression to castration-resistant prostate cancer (CRPC) frequently occurs. Heat shock protein (Hsp) 90 inhibition is a rational therapeutic strategy for CRPC that targets key proteins such as androgen receptor (AR) and protein kinase B (Akt); however, most Hsp90 inhibitors trigger elevation of stress proteins like Hsp27 that confer tumor cell survival and treatment resistance. We hypothesized that cotargeting the cytoprotective chaperone Hsp27 and Hsp90 would amplify endoplasmic reticulum (ER) stress and treatment-induced cell death in cancer. Inducible and constitutive Hsp27 and other HSPs were measured by real-time reverse transcription-polymerase chain reaction and immunoblot assays. The combinations of OGX-427 with Hsp90 inhibitors were evaluated in vitro for LNCaP cell growth and apoptosis and in vivo in CRPC LNCaP xenograft models. Tumor volumes were compared using the Kruskal-Wallis test. Overall survival was analyzed using Kaplan-Meier curves, and statistical significance was assessed with the log-rank test. Hsp90 inhibitors induced expression of HSPs in tumor cells and tissues in a dose- and time-dependent manner; in particular, Hsp27 mRNA and protein levels increased threefold. In vitro, OGX-427 synergistically enhanced Hsp90 inhibitor-induced suppression of cell growth and induced apoptosis by 60% as measured by increased sub-G1 fraction and poly(ADP-ribose) polymerase cleavage. These biologic events were accompanied by decreased expression of HSPs, Akt, AR, and prostate-specific antigen, and induction of ER stress markers (cleaved activating transcription factor 6, glucose-regulated protein 78, and DNA-damage-inducible transcript 3). In vivo, OGX-427 potentiated the anticancer effects of Hsp90 inhibitor PF-04929113 (orally, 25mg/kg) to inhibit tumor growth and prolong survival in CRPC LNCaP xenografts. HSP90 inhibitor-mediated induction of Hsp27 expression can be attenuated by OGX-427, resulting in increased ER stress and apoptosis, and synergistic inhibition of CRPC tumor growth. This study supports the development of targeted strategies using OGX-427 in combination with Hsp90 inhibitors to improve patient outcome in CRPC.