Merck
Indoxyl sulfate promotes cardiac fibrosis with enhanced oxidative stress in hypertensive rats.
Life sciences (2013-05-25)
Maimaiti Yisireyili, Hidehisa Shimizu, Shinichi Saito, Atsushi Enomoto, Fuyuhiko Nishijima, Toshimitsu Niwa
RÉSUMÉ
Cardiovascular disease (CVD) is common in chronic kidney disease (CKD) patients. Indoxyl sulfate (IS) is a nephrovascular uremic toxin that induces oxidative stress in kidney and vascular system. The present study aimed to examine the effect of IS on fibrosis and oxidative stress in rat heart. The effects of IS on heart were examined by Masson's trichrome (MT) staining and immunohistochemistry using: (1) Dahl salt-resistant normotensive rats (DN), (2) Dahl salt-resistant normotensive IS-administered rats (DN+IS), (3) Dahl salt-sensitive hypertensive rats (DH), and (4) Dahl salt-sensitive hypertensive IS-administered rats (DH+IS). DH+IS rats showed significantly increased heart weight and left ventricle weight compared with DN. DH and DH+IS rats showed significantly increased diameter of cardiomyocytes, increased MT-positive fibrotic area, increased staining for transforming growth factor (TGF)-β1, α-smooth muscle actin (SMA), type 1 collagen, NADPH oxidase Nox 4, malondialdehyde (MDA), and 8-hydroxydeoxyguanosine (8-OHdG) and decreased staining for nuclear factor (erythroid-derived 2)-like 2 (Nrf2) and heme oxygenase-1 (HO-1) in the heart compared with DN. More notably, DH+IS rats showed significantly increased diameter of cardiomyocytes, increased fibrotic area, increased staining for TGF-β1, SMA, type 1 collagen, Nox4, 8-OHdG and MDA, and decreased staining for Nrf2 and HO-1 in the heart compared with DH. IS aggravates cardiac fibrosis and cardiomyocyte hypertrophy with enhanced oxidative stress and reduced anti-oxidative defense in hypertensive rats.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Indoxyl sulfate potassium salt