Merck
Dopant-Free Hole Transport Materials with a Long Alkyl Chain for Stable Perovskite Solar Cells.
Nanomaterials (Basel, Switzerland) (2019-07-03)
Kai Wang, Haoran Chen, Tingting Niu, Shan Wang, Xiao Guo, Hong Wang
RÉSUMÉ
Hole transport materials are indispensable to high efficiency perovskite solar cells. Two new hole transporting materials (HTMs), named 4,4'-(9-nonyl-9H-carbazole-3,6-diyl)bis (N,N-bis(4-methoxyphenyl)aniline) (CZTPA-1) and 4,4'-(9-methyl-9H-carbazole-3,6-diyl)bis (N,N-bis(4-methoxyphenyl)aniline)(CZTPA-2), were developed by different alkyl substitution methods. The two compounds, containing a carbazole core and triphenylamine (TPA) groups with different lengths of the alkyl chain, were designed and synthesized through a two-step synthesis approach. The power conversion efficiency (PCE) was found to be affected by the length of the alkyl chain, reaching 7% for CZTPA-1 and 11% for CZTPA-2. Furthermore, the CZTPA-2 still maintained 89.7% of its original performance after 400 h. The proposed results demonstrate the effect of carbon chain substituents on the efficiency of perovskite solar cells (PSCs).