Merck
Mitochondrial protein-induced stress triggers a global adaptive transcriptional programme.
Nature cell biology (2019-03-20)
Felix Boos, Lena Krämer, Carina Groh, Ferris Jung, Per Haberkant, Frank Stein, Florian Wollweber, Adrian Gackstatter, Eva Zöller, Martin van der Laan, Mikhail M Savitski, Vladimir Benes, Johannes M Herrmann
RÉSUMÉ
The cytosolic accumulation of mitochondrial precursors is hazardous to cellular fitness and is associated with a number of diseases. However, it is not observed under physiological conditions. Individual mechanisms that allow cells to avoid cytosolic accumulation of mitochondrial precursors have recently been discovered, but their interplay and regulation remain elusive. Here, we show that cells rapidly launch a global transcriptional programme to restore cellular proteostasis after induction of a 'clogger' protein that reduces the number of available mitochondrial import sites. Cells upregulate the protein folding and proteolytic systems in the cytosol and downregulate both the cytosolic translation machinery and many mitochondrial metabolic enzymes, presumably to relieve the workload of the overstrained mitochondrial import system. We show that this transcriptional remodelling is a combination of a 'wideband' core response regulated by the transcription factors Hsf1 and Rpn4 and a unique mitoprotein-induced downregulation of the oxidative phosphorylation components, mediated by an inactivation of the HAP complex.