Merck
Dihydromyricetin attenuates hypertrophic scar formation by targeting activin receptor-like kinase 5.
European journal of pharmacology (2019-02-27)
Xiaolu Ye, Zhiying Pang, Ningwen Zhu
RÉSUMÉ
Hypertrophic scar (HPS) is a manifestation of abnormal tissue repair, representing excessive extracellular matrix production and abnormal function of fibroblasts, for which no satisfactory treatment is available at present. Here we identified a natural product of flavonoid, dihydromyricetin, could effectively attenuate HPS formation. We showed that local intradermal injection of dihydromyricetin (50 μM) reduced the gross scar area, cross-sectional size of the scar and the scar elevation index in a mechanical load-induced mouse model. In addition, dihydromyricetin treatment also markedly decreased collagen density of the scar tissue. Furthermore, both in vitro and in vivo study both demonstrated that dihydromyricetin inhibited the proliferation, activation, contractile and migration abilities of hypertrophic scar-derived fibroblasts (HSFs) but did not affect HSFs apoptosis. Western blot analysis revealed that dihydromyricetin could down-regulate the phosphorylation of Smad2 and Smad3 of TGF-β signaling. Such bioactivity of dihydromyricetin may result from its selective binding to the catalytic region of activin receptor-like kinase 5 (ALK5), as suggested by the molecular docking study and kinase binding assay (12.26 μM). Above all, dihydromyricetin may prove to be a promising agent for the treatment of HPS and other fibroproliferative disorders.