Merck
Proteomic and functional analysis of Argonaute-containing mRNA-protein complexes in human cells.
EMBO reports (2007-10-13)
Julia Höck, Lasse Weinmann, Christine Ender, Sabine Rüdel, Elisabeth Kremmer, Monika Raabe, Henning Urlaub, Gunter Meister
RÉSUMÉ
Members of the Argonaute (Ago) protein family associate with small RNAs and have important roles in RNA silencing. Here, we analysed Ago1- and Ago2-containing protein complexes in human cells. Separation of Ago-associated messenger ribonucleoproteins (mRNPs) showed that Ago1 and Ago2 reside in three complexes with distinct Dicer and RNA-induced silencing complex activities. A comprehensive proteomic analysis of Ago-containing mRNPs identified a large number of proteins involved in RNA metabolism. By using co-immunoprecipitation experiments followed by RNase treatment, we biochemically mapped interactions within Ago mRNPs. Using reporter assays and knockdown experiments, we showed that the putative RNA-binding protein RBM4 is required for microRNA-guided gene regulation.