Merck
The physical and biochemical effects of pre-rigor high pressure processing of beef.
Meat science (2018-05-12)
James D Morton, Hannah Y-Y Lee, R Grant Pearson, Roy Bickerstaffe
RÉSUMÉ
High pressure processing (HPP) of pre-rigor longissimus thoracis (strip loin) from prime and bull animals substantially decreased the shear force and improved consumer eating attributes of the final meat product. The improved tenderness in both prime and bull meat was associated with a lower myofibrillar fragmentation index and reduced calpain 1 activity which indicated the mechanism of tenderisation was different from that which occurred in chill aged meat. Light microscopy showed disruption to the fibre packing within the muscle and electron microscopy confirmed significant disruption of the Z discs and M lines and disappearance of the A lines. Thus, HPP is associated with a reduction in the structural integrity and strength of the sarcomeres. These effects were consistent in strip loins sourced from prime and bull stock. HPP also led to the movement of glycogen phosphorylase from the sarcoplasmic fraction to the insoluble myofibrillar fraction in all animals and this was associated with a higher pH at 24 h.