Merck
  • Protection of dopaminergic cells by urate requires its accumulation in astrocytes.
Protection of dopaminergic cells by urate requires its accumulation in astrocytes.
Journal of neurochemistry (2012-06-08)
Sara Cipriani, Cody A Desjardins, Thomas C Burdett, Yuehang Xu, Kui Xu, Michael A Schwarzschild
ABSTRACT
Urate is the end product of purine metabolism and a major antioxidant circulating in humans. Recent data link higher levels of urate with a reduced risk of developing Parkinson's disease and with a slower rate of its progression. In this study, we investigated the role of astrocytes in urate-induced protection of dopaminergic cells in a cellular model of Parkinson's disease. In mixed cultures of dopaminergic cells and astrocytes oxidative stress-induced cell death and protein damage were reduced by urate. By contrast, urate was not protective in pure dopaminergic cell cultures. Physical contact between dopaminergic cells and astrocytes was not required for astrocyte-dependent rescue as shown by conditioned medium experiments. Urate accumulation in dopaminergic cells and astrocytes was blocked by pharmacological inhibitors of urate transporters expressed differentially in these cells. The ability of a urate transport blocker to prevent urate accumulation into astroglial (but not dopaminergic) cells predicted its ability to prevent dopaminergic cell death. Transgenic expression of uricase reduced urate accumulation in astrocytes and attenuated the protective influence of urate on dopaminergic cells. These data indicate that urate might act within astrocytes to trigger release of molecule(s) that are protective for dopaminergic cells.
Sigma-Aldrich
Uricase from Arthrobacter globiformis, lyophilized powder, 15-30 units/mg protein (biuret)
Sigma-Aldrich
Uricase from Bacillus fastidiosus, lyophilized, ~9 U/mg
Sigma-Aldrich
Uricase from Candida sp., recombinant, expressed in E. coli, lyophilized powder, ≥2 units/mg solid