Merck
  • Emerging Approaches to Investigate the Influence of Transition Metals in the Proteinopathies.
Emerging Approaches to Investigate the Influence of Transition Metals in the Proteinopathies.
Cells (2019-10-30)
Frederik Lermyte, James Everett, Jake Brooks, Francesca Bellingeri, Kharmen Billimoria, Peter J Sadler, Peter B O'Connor, Neil D Telling, Joanna F Collingwood
ABSTRACT
Transition metals have essential roles in brain structure and function, and are associated with pathological processes in neurodegenerative disorders classed as proteinopathies. Synchrotron X-ray techniques, coupled with ultrahigh-resolution mass spectrometry, have been applied to study iron and copper interactions with amyloid β (1-42) or α-synuclein. Ex vivo tissue and in vitro systems were investigated, showing the capability to identify metal oxidation states, probe local chemical environments, and localize metal-peptide binding sites. Synchrotron experiments showed that the chemical reduction of ferric (Fe3+) iron and cupric (Cu2+) copper can occur in vitro after incubating each metal in the presence of Aβ for one week, and to a lesser extent for ferric iron incubated with α-syn. Nanoscale chemical speciation mapping of Aβ-Fe complexes revealed a spatial heterogeneity in chemical reduction of iron within individual aggregates. Mass spectrometry allowed the determination of the highest-affinity binding region in all four metal-biomolecule complexes. Iron and copper were coordinated by the same N-terminal region of Aβ, likely through histidine residues. Fe3+ bound to a C-terminal region of α-syn, rich in aspartic and glutamic acid residues, and Cu2+ to the N-terminal region of α-syn. Elucidating the biochemistry of these metal-biomolecule complexes and identifying drivers of chemical reduction processes for which there is evidence ex-vivo, are critical to the advanced understanding of disease aetiology.
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium citrate tribasic, ≥97% (titration)
Sigma-Aldrich
1,10-Phenanthroline, ≥99%