Skip to Content
Merck
All Photos(2)

Key Documents

V900395

Sigma-Aldrich

trans-4-Hydroxy-L-proline

Vetec, reagent grade, 99%

Synonym(s):

(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
Beilstein:
81441
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

crystalline powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

273 °C (dec.) (lit.)

SMILES string

O[C@H]1CN[C@@H](C1)C(O)=O

InChI

1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1

InChI key

PMMYEEVYMWASQN-DMTCNVIQSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Natural constituent of animal structural proteins such as collagen and elastin.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahrzad Hosseininia et al.
BMC musculoskeletal disorders, 14, 18-18 (2013-01-11)
It remains to be shown whether OA shares molecular similarities between different joints in humans. This study provides evidence for similarities in cartilage molecular damage in osteoarthritic (OA) joints. Articular cartilage from osteoarthritic hip joints were analysed and compared to
Jacob Kofoed et al.
Chemical communications (Cambridge, England), (14)(14), 1482-1484 (2006-04-01)
The aldol reaction of acetone with aldehydes in aqueous medium under catalysis by zinc-proline (Zn(L-Pro)2) and secondary amines such as proline, (2S,4R)-4-hydroxyproline (Hyp) and (S)-(+)-1-(2-pyrrolidinomethyl)pyrrolidine (PMP) is shown to proceed by an enamine mechanism, as evidenced by reductive trapping of
Mari Ogawa-Ohnishi et al.
Nature chemical biology, 9(11), 726-730 (2013-09-17)
Hydroxyproline (Hyp) O-arabinosylation is a post-translational modification that is prominent in extracellular glycoproteins in plants. Hyp O-arabinosylation is generally found in these glycoproteins in the form of linear oligoarabinoside chains and has a key role in their function by contributing
Massimo Petrera et al.
Arthroscopy : the journal of arthroscopic & related surgery : official publication of the Arthroscopy Association of North America and the International Arthroscopy Association, 29(10), 1685-1692 (2013-10-01)
This study aimed to determine the effects of platelet-rich plasma (PRP) on the histologic, biochemical, and biomechanical properties of tissue-engineered cartilage. Chondrocytes isolated from bovine metacarpal-phalangeal articular cartilage were seeded on top of a porous ceramic substrate (calcium polyphosphate [CPP]).
Ville V Meretoja et al.
Biomaterials, 34(17), 4266-4273 (2013-03-16)
In this work, we investigated the effects of lowered oxygen tension (20% and 5% O2) on the chondrogenesis and hypertrophy of articular chondrocytes (ACs), mesenchymal stem cells (MSCs) and their co-cultures with a 30:70 AC:MSC ratio. Cells were cultured for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service