Skip to Content
Merck
All Photos(1)

Key Documents

V900199

Sigma-Aldrich

Sodium silicate solution

Vetec, reagent grade

Synonym(s):

Sodium trisilicate solution, Water glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O(SiO2)x · xH2O
UNSPSC Code:
12352300

grade

reagent grade

product line

Vetec

form

liquid

composition

NaOH, 11.4-14.4% titration (by HCl)
Si, 11.0-15.0% gravimetric

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service