Skip to Content
Merck
All Photos(1)

Key Documents

442311

Supelco

2,4-Diaminotoluene

analytical standard

Synonym(s):

4-Methyl-m-phenylenediamine, 2,4-Diaminotoluene, 2,4-Toluenediamine, 4-Methyl-1,3-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)2
CAS Number:
Molecular Weight:
122.17
Beilstein:
2205839
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

283-285 °C (lit.)

mp

97-99 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Cc1ccc(N)cc1N

InChI

1S/C7H10N2/c1-5-2-3-6(8)4-7(5)9/h2-4H,8-9H2,1H3

InChI key

VOZKAJLKRJDJLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Diaminotoluene is a primary aromatic amine (PAA) that is used in industrial and agricultural applications. It is employed as an intermediate in the manufacture of toluene diisocyanate and polyurethane foam, elastomers and coatings.
2,4-Diaminotoluene is an organic compound, which is used for the production of industrial dyes.

Application

2,4-Diaminotoluene may be used as a reference standard in the determination of 2,4-diaminotoluene in biological samples using high-performance liquid chromatography (HPLC).
2,4-Diaminotoluene may be used as an analytical reference standard for the determination of the analyte in food packing materials, textile samples and hair dyes by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The analysis of arylamines and phenols in oxidation-type hair dyes by paper chromatography
Smyth RB and McKeown GG
Journal of Chromatography A, 16(1), 454-459 (1964)
Determination of primary aromatic amines in water food simulant using solid-phase analytical derivatization followed by gas chromatography coupled with mass spectrometry
Brede C, et al.
Journal of Chromatography A, 983(1-2), 35-42 (2003)
High-performance liquid chromatography of 2, 6-and 2, 4-diaminotoluene, and its application to the determination of 2, 4-diaminotoluene in urine and plasma.
Unger DP and Friedman AM
Journal of Chromatography A, 174(2), 379-384 (1979)
Determination of hemoglobin adduct levels of the carcinogen 2, 4-diaminotoluene using gas chromatography-electron impact positive-ion mass spectrometry
Wilson PM, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 667(1), 166-172 (1995)
LC-MS/MS method for the confirmatory determination of aromatic amines and its application in textile analysis.
Sutthivaiyakit P, et al.
Analytical and Bioanalytical Chemistry, 381(1), 268-276 (2005)

Protocols

GC Analysis of Anilines on Equity®-5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service