13094-U
Glass Packed GC Column (fits Agilent, Configuration "A")
phase none, matrix 80/100 Porapak QS support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
About This Item
Recommended Products
material
glass column
manufacturer/tradename
Agilent
parameter
0-250 °C temperature (isothermal or programmed)
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
matrix
80/100 Porapak QS support
matrix active group
none phase
column type
packed GC
compatibility
for use with Agilent Configuration A
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service