Skip to Content
Merck
All Photos(1)

Key Documents

T0688

Sigma-Aldrich

Anti-Transforming Growth Factor-β1 antibody produced in chicken

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-TGF-β1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
clone:
polyclonal
application:
Neutral
WB
species reactivity:
human
technique(s):
neutralization: suitable
western blot: suitable
citations:
6

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGFB1(7040)

Related Categories

General description

Transforming Growth Factor-β1 (TGF β1) is a multifunctional protein that can stimulate or inhibit cellular proliferation. Mesenchymal origin cells are stimulated by TGF β1 while T and B lymphocytes, keratinocytes, hepatocytes and epithelial cells are inhibited by TGF β1. It facilitates in tissue growth and differentiation by participating in myogenesis, osteogenesis, adipogenesis, chondrogenesis, epithelial cell differentiation and immune cell function. Anti-Transforming Growth Factor-β1 antibody can be used in neutralization assay. Chicken anti-TGF β1 reacts specifically with recombinant human TGF-β1 or TGF-β1.2.

Specificity

The antibody will neutralize the biological activity of recombinant human TGF-β1 and porcine TGF-β1.2.

Immunogen

Recombinant human TGF-β1 expressed in CHO cells.

Application

Anti-Transforming Growth Factor-β1 antibody can be used in western blotting.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Kehrl et al.
The Journal of experimental medicine, 163(5), 1037-1050 (1986-05-01)
This study examines the potential role of transforming growth factor beta (TGF-beta) in the regulation of human T lymphocyte proliferation, and proposes that TGF-beta is an important autoregulatory lymphokine that limits T lymphocyte clonal expansion, and that TGF-beta production by
G D Shipley et al.
Cancer research, 46(4 Pt 2), 2068-2071 (1986-04-01)
Type beta transforming growth factor-growth inhibitor (TGF beta/GI) causes normal human prokeratinocytes to arrest growth predominantly in the G1 phase of the cell cycle within 48 h after log phase cultures are exposed to the factor in serum-free medium. The
S Cheifetz et al.
Cell, 48(3), 409-415 (1987-02-13)
A new homodimer form of transforming growth factor-beta (TGF-beta), TGF-beta 2, has been identified in porcine blood platelets. TGF-beta 2 is homologous to ordinary TGF-beta (TGF-beta 1), which is also present in platelets. TGF-beta 1.2, a heterodimer containing one TGF-beta
C B Childs et al.
Proceedings of the National Academy of Sciences of the United States of America, 79(17), 5312-5316 (1982-09-01)
High concentrations of fetal bovine serum induced colony formation in soft agar by anchorage-dependent, nontransformed mouse AKR-2B and rat NRK cells. The colony-stimulating activity in fetal bovine serum was precipitated by 45% saturated ammonium sulfate and migrated in molecular sieve
I Hayashi et al.
Journal of cellular physiology, 125(1), 82-90 (1985-10-01)
Primary monolayer cultures of adult rat hepatocytes can be induced to undergo DNA synthesis in serum-free medium in the presence of insulin, glucagon, and epidermal growth factor (three factors). We have found that hepatocyte DNA synthesis is affected not only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service