Skip to Content
Merck
All Photos(2)

Key Documents

SRP6546

Sigma-Aldrich

Procathepsin L human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

CTSL, CTSL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

39.4 kDa (18-333 aa + N-terminal poly-his tag.)

packaging

pkg of 1 mg
pkg of 100 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−70°C

Gene Information

human ... CTSL1(1514)

General description

Cathepsin L is a lysosomal protease that can be secreted from the cell and is implicated to play a role in protein degradation, arthritis, apoptosis, and cancer. TheProcathepsin L can be auto-catalytically activated to mature Cathepsin L (~24 kDa) by adding an equal volume of 0.2 M NaOAc, 5 mM EDTA pH 4.0 containing fresh 5 mM DTT at RT. The enzyme is fully activated within 30 min of incubation at RT. After activation mature Cathepsin L is highly auto-proteolytic at pH 4.0, and care must be taken to avoid self-proteolysis. If the activated enzyme is not used immediately, methylmethanthiosulfonate (1mM final concentration; MeS-SO2Me; MMTS) could be added and sample should be frozen at -80°C. The activity can be then restored by adding L-cysteine (3 M excess over MMTS) to the enzyme solution.

Physical form

1 mg/mL in 25 mM Na2HPO4, 500 mM NaCl, pH 7.0.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service