Skip to Content
Merck
All Photos(1)

Key Documents

SRP6425

Sigma-Aldrich

SERPIN A3 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

AACT, ACT, GIG24, GIG25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 46 kDa
observed mol wt 55-70 kDa (DTT-reduced. Protein migrates due to glycosylation. His 23 is the predicted N-terminal.)

packaging

pkg of 10 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SERPINA3(12)

General description

Serpin A3 is also known as α-1-antichymotrypsin (ACT or AACT) gene with five exons and four introns spanning 12.8kb long, is mapped to human chromosome 14q32.13. The encoded protein is a 423 amino acid with the molecular weight of ~ 55-66kDa and it belongs to the family of serpin superfamily of protease inhibitors. Serpin A3 consists of three β sheets, eight α helices and an active site situated within a hypervariable reactive center loop (RCL).

Biochem/physiol Actions

Overexpression of SERPIN A3 leads to endometrial carcinoma (EC), Therefore targeting SERPIN A3 can be a potent therapeutic method for EC. In addition, elevated expression of SERPIN A3 expression has been observed in various types of carcinomas including breast carcinoma, hepatocellular carcinoma, prostate carcinoma and adenocarcinoma of the lung. SERPINA3 is associated with the pathogenesis of chronic obstructive pulmonary disease (COPD), Parkinson′s disease (PD), Alzheimer′s disease (AD), Stroke, Cystic Fibrosis, Cerebral Haemorrhage and Multiple System Atrophy. SERPIN A3 inhibits the activity of pancreatic chymotrypsin, leukocyte, cathepsin G, mast cell chymases, human glandular, kallikrein 2, kallikrein 3 (prostate specific antigen), pancreatic cationic elastase and an uncharacterized lung serum protease. SERPIN A3 can bind to DNA, this property is unique among the members of SERPIN family.

Physical form

Lyophilized from 0.22 μm filtered solution in 20 mM HEPES, 150 mM NaCl, pH 7.5. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Down-regulated expression of SERPIN genes located on chromosome 18q21 in oral squamous cell carcinomas.
Shiiba M
Oncology Reports, 24(1), 241-249 (2010)
Crystal Baker et al.
Frontiers in bioscience : a journal and virtual library, 12, 2821-2835 (2007-05-09)
Recent advances in proteomic, transcriptomic and genomic technologies have revealed much about the ACT protein and gene. In this review, we summarize our current understanding of the structure and potential physiological roles of the ACT protein, catalogue the regulatory elements
SERPINA3 promotes endometrial cancer cells growth by regulating G2/M cell cycle checkpoint and apoptosis.
Yang GD
International Journal of Clinical and Experimental Pathology, 7(4), 1348-1358 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service