Skip to Content
Merck
All Photos(1)

Key Documents

SRP6282

Sigma-Aldrich

Nicotinamide N-Methyltransferase (hNNMT) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE)

Synonym(s):

NNMT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

liquid

mol wt

31.7 kDa (263 aa + NT 6xHis-tag)

packaging

pkg of 1 mg
pkg of 100 μg
pkg of 20 μg

concentration

3.42 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NNMT(4837)

General description

N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. NNMT is an important cytosolic methyltransferase catalyzing the N-methylation of nicotinamide, pyridines and structural analogs, playing a crucial role in the biotransformation and detoxification of many xenobiotic compounds. In the N-methylation process, NNMT uses S-adenosyl methionine as the methyl donor and nicotinamide as methyl acceptor. NNMT is mostly expressed in the liver, and a lower expression is seen in the kidney, lung, skeletal muscle, placenta and heart. It may also play an important role in regulating biological processes related to N-methyl nicotinamide such as anti-inflammatory, anti-thrombotic, vasoprotective, and gastroprotective properties. Increased NNMT activity was reported in many kinds of tumors. NNMT is a potential biomarker and therapeutic target in caner diagnosis and treatment. NNMT serum levels have significance in the premature detection and in the management of patients with colorectal cancer. The recombinant protein includes hNNMT with N-terminal His-tag.

Physical form

3.42 mg/mL solution in 25 mM Tris (pH8.0) buffer containing 20% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service