Skip to Content
Merck
All Photos(1)

Key Documents

SRP5180

Sigma-Aldrich

FADD, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

GIG3, MGC8528, MORT1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~27 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FADD(8772)

General description

FADD or Fas-Associated protein with Death Domain is an adaptor molecule that mediates death signaling by the Fas-receptor, tumor necrosis factor receptor and TRAIL-receptor. FADD binds to these receptors via the C-terminus Death Domain which then unmasks the N-terminal effector domain of FADD thereby allowing it to recruit caspase-8 and activate the cysteine protease cascade leading to apoptosis. Cells lacking FADD are defective in intracellular double-stranded RNA (dsRNA)-activated gene expression, including production of type I (alpha/beta) interferons and are thus very susceptible to viral infection.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Saeed Sheikh et al.
Cell cycle (Georgetown, Tex.), 2(4), 346-347 (2003-07-10)
Fas-associated death domain protein (FADD) is an adaptor molecule that bridges the interactions between membrane death receptors and initiator caspases. Thus, the site of its action has always been expected to be the cytoplasmic death-inducing signaling complex (DISC). Recent evidence
Siddharth Balachandran et al.
Nature, 432(7015), 401-405 (2004-11-19)
Vertebrate innate immunity provides a first line of defence against pathogens such as viruses and bacteria. Viral infection activates a potent innate immune response, which can be triggered by double-stranded (ds)RNA produced during viral replication. Here, we report that mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service