Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500053

Sigma-Aldrich

Anti-MIPU1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Myocardial ischemic preconditioning upregulated protein 1, Anti-ZNF667, Anti-Zinc finger protein 667

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF667(63934)

Immunogen

MIPU1 antibody was raised against a 17 amino acid peptide near the amino terminus of human MIPU1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Mipu1, also known as zinc finger protein 667 or ZNF667, encodes a nuclear-localized protein containing 14 carboxy-terminal zinc finger motifs and an amino-terminal KRAB domain. This protein is highly expressed in heart and brain and is upregulated in rat heart after a transient ischemia-reperfusion procedure. Overexpression experiments suggest that Mipu1 suppresses the transcriptional activities of AP-1 and SRE in the MAPK signaling pathway and thus may play a role in the pathogenesis of cardiac and vascular disease. At least four isoforms of MIPU1 are known to exist.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500053.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service