Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501449

Sigma-Aldrich

Anti-SEPT4 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ARTS, Anti-BRADEION, Anti-CE5B3, Anti-H5, Septin 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, bovine, canine

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEPT4(5414)

Related Categories

Immunogen

Peptide with sequence RNKLTRESGTD, from the internal region of the protein sequence according to NP_004565.1; NP_536341.1, NP_001185642.1

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jozsef Maléth et al.
Nature communications, 5, 5843-5843 (2014-12-18)
The Orai1-STIM1 current undergoes slow Ca(2+)-dependent inactivation (SCDI) mediated by the binding of SARAF to STIM1. Here we report the use of SCDI by SARAF as a probe of the conformation and microdomain localization of the Orai1-STIM1 complex. We find

Global Trade Item Number

SKUGTIN
SAB2501449-100UG4061831450897

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service