Skip to Content
Merck
All Photos(3)

Key Documents

SAB1411266

Sigma-Aldrich

Anti-SGTA antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

SGT, alphaSGT, hSGT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 34.1 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGTA(6449)

General description

This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function. (provided by RefSeq)

Immunogen

SGTA (NP_003012.1, 1 a.a. ~ 313 a.a) full-length human protein.

Sequence
MDNKKRLAYAIIQFLHDQLRHGGLSSDAQESLEVAIQCLETAFGVTVEDSDLALPQTLPEIFEAAATGKEMPQDLRSPARTPPSEEDSAEAERLKTEGNEQMKVENFEAAVHFYGKAIELNPANAVYFCNRAAAYSKLGNYAGAVQDCERAICIDPAYSKAYGRMGLALSSLNKHVEAVAYYKKALELDPDNETYKSNLKIAELKLREAPSPTGGVGSFDIAGLLNNPGFMSMASNLMNNPQIQQLMSGMISGGNNPLGTPGTSPSQNDLASLIQAGQQFAQQMQQQNPELIEQLRSQIRSRTPSASNDDQQE

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Zhu et al.
Journal of molecular histology, 45(6), 665-677 (2014-07-17)
Small glutamine-rich tetratricopeptide repeat-containing protein alpha (SGTA) was reported to be implicated in various cellular processes and involved in control of cell cycle regulation and transcription. It may play a critical role in oncogenesis. In this study, to investigate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service