Skip to Content
Merck
All Photos(2)

Key Documents

SAB1407998

Sigma-Aldrich

Anti-PHF17 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ22479, JADE1, KIAA1807

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~58.4 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF17(79960)

Related Categories

General description

Mouse polyclonal antibody raised against a full-length human PHF17 protein.
PHF17 (PHD finger protein 17) is a short-lived, kidney-enriched Jade protein consisting of canonical plant homeodomain (PHD) finger protein and a non-canonical extended PHD with zinc-binding ability. It is localized in the nucleus and is highly expressed in the kidney and renal proximal tubule cells.

Immunogen

PHF17 (NP_079176.2, 1 a.a. ~ 509 a.a) full-length human protein.

Sequence
MKRGRLPSSSEDSDDNGSLSTTWSQNSRSQHRRSSCSRHEDRKPSEVFRTDLITAMKLHDSYQLNPDEYYVLADPWRQEWEKGVQVPVSPGTIPQPVARVVSEEKSLMFIRPKKYIVSSGSEPPELGYVDIRTLADSVCRYDLNDMDAAWLELTNEEFKEMGMPELDEYTMERVLEEFEQRCYDNMNHAIETEEGLGIEYDEDVVCDVCQSPDGEDGNEMVFCDKCNICVHQACYGILKVPEGSWLCRTCALGVQPKCLLCPKKGGAMKPTRSGTKWVHVSCALWIPEVSIGSPEKMEPITKVSHIPSSRWALVCSLCNEKFGASIQCSVKNCRTAFHVTCAFDRGLEMKTILAENDEVKFKSYCPKHSSHRKPEESLGKGAAQENGAPECSPRNPLEPFASLEQNREEAHRVSVRKQKLQQLEDEFYTFVNLLDVARALRLPEEVVDFLYQYWKLKRKVNFNKPLITPKKDEEDNLAKREQDVLFRRLQLFTHLRQDLERVMIDTDTL

Application

Anti-PHF17 antibody produced in mouse is suitable for indirect immunofluorescence and western blot analysis.

Biochem/physiol Actions

PHF17 (PHD finger protein 17) is associated with several cellular activities such as chromatin remodeling, renal tubular epithelial cell differentiation, growth suppression, apoptosis and protein-protein interactions. It possesses transcriptional and endogenous histone acetyltransferase (HAT) activity. It acts as a transcriptional co-activator in the TIP60 mediated histone H4/H2A specific HAT activity. It has been reported that PHF17 may play a role in the renal cancer and von Hippel-Lindau disease. PHF17 also possesses tumour suppressor property. In the renal tumorigenesis, it controls canonical Wnt signaling pathway by ubiquitylating oncoprotein β-catenin in Wnt-responsive manner.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria V Panchenko et al.
The Journal of biological chemistry, 279(53), 56032-56041 (2004-10-27)
Jade-1 was identified as a protein partner of the von Hippel-Lindau tumor suppressor pVHL. The interaction of Jade-1 and pVHL correlates with renal cancer risk. We have investigated the molecular function of Jade-1. Jade-1 has two zinc finger motifs called
Vipul C Chitalia et al.
Nature cell biology, 10(10), 1208-1216 (2008-09-23)
The von Hippel-Lindau protein pVHL suppresses renal tumorigenesis in part by promoting the degradation of hypoxia-inducible HIF-alpha transcription factors; additional mechanisms have been proposed. pVHL also stabilizes the plant homeodomain protein Jade-1, which is a candidate renal tumour suppressor that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service