Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402269

Sigma-Aldrich

Monoclonal Anti-MAP3K3 antibody produced in mouse

clone 1H3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MAPKKK3, MEKK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~35.9 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP3K3(4215)

General description

This gene product is a 626-amino acid polypeptide that is 96.5% identical to mouse Mekk3. Its catalytic domain is closely related to those of several other kinases, including mouse Mekk2, tobacco NPK, and yeast Ste11. Northern blot analysis revealed a 4.6-kb transcript that appears to be ubiquitously expressed. This protein directly regulates the stress-activated protein kinase (SAPK) and extracellular signal-regulated protein kinase (ERK) pathways by activating SEK and MEK1/2 respectively; it does not regulate the p38 pathway. In cotransfection assays, it enhanced transcription from a nuclear factor kappa-B (NFKB)-dependent reporter gene, consistent with a role in the SAPK pathway. Alternatively spliced transcript variants encoding distinct isoforms have been observed. (provided by RefSeq)

Immunogen

MAP3K3 (AAH10464.1, 1 a.a. ~ 89 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MNEANVMLPYSGKEEPVLPVAMTLPLPGRGPRCGTAATEGGSSFVNAVVSVLQVGVTLMLYPVSKLETVCALWALSTPALGLGLGCIEK

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Quan Cao et al.
Asian Pacific journal of cancer prevention : APJCP, 15(13), 5271-5276 (2014-07-22)
Mitogen-activated protein kinase/extracellular signal-regulated kinase kinase kinase 3 (MEKK3) is an important protein kinase and a member of the MAPK family, which regulates cellular responses to environmental stress and serves as key integration points along the signal transduction cascade that
Nayara I Viana et al.
The International journal of biological markers, 29(3), e246-e252 (2014-01-30)
The aim of this study was to analyze the roles of miR-143 and miR-145, as well as the gene and protein expression of their targets (KRAS, ERK5, MAP3K3, and MAP4K4) in the pathogenesis of benign prostatic hyperplasia (BPH). We analyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service