Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300317

Sigma-Aldrich

Anti-LRP6 (N-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-LDL receptor-related protein 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate:
unconjugated
application:
ELISA (i)
clone:
polyclonal
species reactivity:
human
citations:
2
technique(s):
indirect ELISA: 1:1000
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

General description

Low density lipoprotein (LDL) receptor-related protein (LRP), a member of the LDL receptor family, binds multiple classes of ligands and has been implicated in a broad range of normal and disease processes involving lipid metabolism, protease clearance, and cell migration. Structurally, members of the LDLR family share homology within their extracellular domains, which are highlighted by the presence of clusters of ligand-binding repeats. LRP is a large endocytic receptor that participates in several biological pathways and plays prominent roles in lipoprotein metabolism and in the catabolism of proteinases involved in coagulation and fibrinolysis. LRP also mediates the cellular entry of certain viruses and toxins and facilitates the activation of various lysosomal enzymes. All LRPs are expressed in the central nervous system and, for most receptors, animal models have shown that they are indispensable for successful neurodevelopment. The mechanisms by which they regulate the formation of the nervous system are varied and include the transduction of extracellular signals and the modulation of intracellular signal propagation, as well as cargo transport, the function most commonly attributed to this gene family.

Immunogen

LRP6 (NP_002327, )
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human LRP6.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine M Willis et al.
PloS one, 9(8), e103966-e103966 (2014-08-05)
Expression of the glycosaminoglycan chondroitin sulfate-E (CS-E) is misregulated in many human cancers, including breast cancer. Cell-surface associated CS-E has been shown to have pro-tumorigenic functions, and pharmacological treatment with exogenous CS-E has been proposed to interfere with tumor progression
Wenyan Lu et al.
Journal of cellular biochemistry, 115(10), 1799-1807 (2014-06-07)
Emerging evidence indicates that activation of Wnt/β-catenin signaling at the cell surface results in inhibition of glycogen synthase kinase 3β (GSK3β), leading to activation of mTORC1 signaling in cancer cells. The low density lipoprotein receptor-related protein-6 (LRP6) is an essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service