Skip to Content
Merck
All Photos(2)

Key Documents

HPA017973

Sigma-Aldrich

Anti-PLEKHG2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ARHGEF42, Anti-CLG, Anti-FLJ00018, Anti-pleckstrin homology domain containing, family G (with RhoGef domain) member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
2
technique(s):
immunohistochemistry: 1:200- 1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

ATRRELFSGSNPGKLGEPPSGGKAGPEEDEEGVSFTDFQPQDVTQHQGFPDELAFRSCSEIRSAWQALEQGQLARPGFPEPLLILEDSDLGGDSGSGKAGAPSSERTASRVRELARLYSERI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLEKHG2(64857)

General description

Pleckstrin homology domain containing family G member 2 (PLEKHG2) is a guanine nucleotide exchange factor for GTPases like Rac and Cdc42. It belongs to the Dbl family.

Immunogen

pleckstrin homology domain containing, family G (with RhoGef domain) member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Pleckstrin homology domain containing family G member 2 (PLEKHG2) plays an important role in signaling pathways which deal with reorganization of the actin skeleton. Activation of the Ras/Mitogen-activated protein kinase (MAPK) pathway leads to the phosphorylation of PLEKHG2 and it further regulates processes controlling changes in cell shape. Studies have shown that it is upregulated in many leukemia cell lines and it is an important factor for lymphocyte migration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72558

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuya Sato et al.
The Journal of biological chemistry, 289(14), 10045-10056 (2014-02-21)
FLJ00018/PLEKHG2 is a guanine nucleotide exchange factor for the small GTPases Rac and Cdc42 and has been shown to mediate the signaling pathways leading to actin cytoskeleton reorganization. The function of FLJ00018 is regulated by the interaction of heterotrimeric GTP-binding
Caitlin Runne et al.
Molecular and cellular biology, 33(21), 4294-4307 (2013-09-05)
PLEKHG2 is a Dbl family Rho guanine nucleotide exchange factor (RhoGEF) whose gene was originally identified as being upregulated in a leukemia mouse model and was later shown to be activated by heterotrimeric G protein βγ (Gβγ) subunits. However, its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service