Skip to Content
Merck
All Photos(1)

Key Documents

GW22473C

Sigma-Aldrich

Anti-PRKCQ antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-22473C

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKCQ(5588)

General description

PRKCQ (protein kinase C θ) is majorly expressed in T-cells, and is a member of the PKC protein family. It belongs to the subfamily of proteins which are induced by diacylglycerol (DAG), rather than Ca2+. It is the only PKC member that is localized to the immunological synapse.

Immunogen

Immunogen Sequence: GI # 5453976, sequence 610-673
Recombinant protein kinase C, theta

Application

Anti-PRKCQ antibody produced in chicken is suitbale for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Protein kinase C θ (PKC-θ) promotes epithelial to mesenchymal transition (EMT) by acting as a critical chromatin-anchored switch for inducible genes. Chromatinized PKC-θ exists as an active transcription complex and is essential to establish a permissive chromatin state at signature EMT genes. It plays a central role in TCR-induced IL-2 production and T-cell proliferation. PKCθ activity is regulated by the intracellular redox state. It is the only T cell-expressed PKC that localizes selectively to the center of the immunological synapse (IS) following conventional T cell antigen stimulation. This unique localization is required for PKCθ-mediated downstream signaling. These attributes of PKCθ makes it an attractive target for catalytic or allosteric inhibitors. PKCθ may be associated with type 2 diabetis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judy L Cannon et al.
PloS one, 8(11), e78940-e78940 (2013-11-20)
Cell motility is a fundamental process crucial for function in many cell types, including T cells. T cell motility is critical for T cell-mediated immune responses, including initiation, activation, and effector function. While many extracellular receptors and cytoskeletal regulators have
Anjum Zafar et al.
Molecular and cellular biology, 34(16), 2961-2980 (2014-06-04)
Epithelial to mesenchymal transition (EMT) is activated during cancer invasion and metastasis, enriches for cancer stem cells (CSCs), and contributes to therapeutic resistance and disease recurrence. Signal transduction kinases play a pivotal role as chromatin-anchored proteins in eukaryotes. Here we
Julia Szendroedi et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(26), 9597-9602 (2014-07-01)
Muscle insulin resistance is a key feature of obesity and type 2 diabetes and is strongly associated with increased intramyocellular lipid content and inflammation. However, the cellular and molecular mechanisms responsible for causing muscle insulin resistance in humans are still
Elizabeth Yan Zhang et al.
Advances in pharmacology (San Diego, Calif.), 66, 267-312 (2013-02-26)
Protein kinase C-theta (PKCθ) is a protein kinase C (PKC) family member expressed predominantly in T lymphocytes, and extensive studies addressing its function have been conducted. PKCθ is the only T cell-expressed PKC that localizes selectively to the center of
Marina Rode von Essen et al.
European journal of immunology, 43(6), 1659-1666 (2013-02-26)
PKC-θ plays a central role in TCR-induced IL-2 production and T-cell proliferation. The aim of the present study was to analyse how PKC-θ is regulated in human T cells during T-cell activation and differentiation. We show that PKC-θ is found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service