Skip to Content
Merck
All Photos(1)

Key Documents

CMT64/61

86082105, mouse lung, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
mouse lung
growth mode:
Adherent
karyotype:
Modal no. 85
morphology:
Epithelial
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

CMT64/61, 86082105

biological source

mouse lung

growth mode

Adherent

karyotype

Modal no. 85

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C57BL/1CRF lung carcinoma

Cell Line Description

Derived from an alveologenic lung carcinoma from a C57BL/1CRF mouse. Can be maintained in a completely defined serum free medium, but the growth rate is much slower than in supplemented serum. Used in studies of lung metastases.

Application

Metastasis studies

Culture Medium

Waymouth′s MB 752/1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service