Skip to Content
Merck
All Photos(1)

Key Documents

AC29 cell line from mouse lung

10092308, mouse lung, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source:
mouse lung
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial-like
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer
Pricing and availability is not currently available.

Product Name

AC29 cell line from mouse lung, 10092308

biological source

mouse lung

description

Malignant mesothelioma from mouse lung

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

Cell Line Origin

Mouse malignant mesothelioma

Cell Line Description

Mice (CBA strain female 6-8 weeks) were exposed to crocidiolite asbestos through intraperitoneal injection resulting in tumour development (both ascites and solid tumours). Cultures were established from malignant mesothelial cells obtained from ascites fluid. Cells are tumourigenic in syngeneic imunocompetent mice. Cells grow as adherent monolayers and are epithelial like. Cell Bank Australia reference number: CBA0152

Application

Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods.

Culture Medium

RPMI1640 (with 2mM L-Glutamine+ 25mM HEPES) + 5% FCS

Subculture Routine

Optimal split ratio 1:8 - 1:16 (seeding density 0.6 x104 cells/cm2).Cells incubated at 37°C with 5% CO2.Harvest the cells using 0.05% Trypsin/EDTA at 37°C for 5 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Richard Lake - University of Western Australia, Australia

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service