Recommended Products
Product Name
H400, 06092006
biological source
human mouth
growth mode
Adherent
karyotype
Not specified
morphology
Polygonal
products
Not specified
receptors
TGFbeta type I receptor, TGFbeta II receptor, TGFbeta III receptor
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
Cell Line Origin
Cell Line Description
Culture Medium
Subculture Routine
Other Notes
Disclaimer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service