Skip to Content
Merck
All Photos(3)

Key Documents

AV36578

Sigma-Aldrich

Anti-ANXA3 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Annexin A3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA3(306)

Immunogen

Synthetic peptide directed towards the N terminal region of human ANXA3

Biochem/physiol Actions

ANXA3 is a calcium-dependent, phospholipid-binding protein that belongs to the annexin family. It is an anticoagulant and promotes tumorigenesis and chemoresistance in hepatocellular carcinoma. ANXA3 inhibits the activity of phospholipase A2 and prevents the cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate.

Sequence

Synthetic peptide located within the following region: MASIWVGHRGTVRDYPDFSPSVDAEAIQKAIRGIGTDEKMLISILTERSN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Wu et al.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico, 15(2), 106-110 (2012-09-27)
Annexin family proteins are a well-known multigene family of Ca(2+)-regulated phospholipid- and membrane-binding proteins. As one of the annexin family genes/proteins, accumulated researches have begun to reveal that annexin A3 (Anxa3) exhibits important roles in tumor development, metastasis and drug
Qiu-Zhong Pan et al.
Molecular carcinogenesis, 54(8), 598-607 (2014-01-01)
Annexin A3 (ANXA3) has been found to play important roles in cancer progression, metastasis, and drug resistance; however, its role in hepatocellular carcinoma (HCC) remains unknown. In this study, we investigated the expression level, clinical significance and biologic function of
Volker Gerke et al.
Nature reviews. Molecular cell biology, 6(6), 449-461 (2005-06-02)
Eukaryotic cells contain various Ca(2+)-effector proteins that mediate cellular responses to changes in intracellular Ca(2+) levels. A unique class of these proteins - annexins - can bind to certain membrane phospholipids in a Ca(2+)-dependent manner, providing a link between Ca(2+)
Hiroshi Nishiura et al.
Experimental and molecular pathology, 97(2), 241-246 (2014-07-19)
The roles of annexin A3 (ANXA3) in macrophages are not fully understood. In contrast to C5a, we have demonstrated that C-terminal ribosomal protein S19 (RP S19)-tagged S-tagged C5a (S-tagged C5a/RP S19) raises an alternative cytoplasmic calcium oscillation by extracellular calcium
Tangli Xiao et al.
Molecular and cellular biochemistry, 394(1-2), 145-154 (2014-05-23)
The aim was to explore the effects of rapamycin on autophagy and injury of podocytes in streptozocin (STZ)-induced type 1 diabetic mice, and its role in delaying progression of diabetic nephropathy. In this study, male Balb/c mice were divided into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service