Skip to Content
Merck
All Photos(1)

Key Documents

36705

Supelco

2,5-Dichlorobenzoic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CO2H
CAS Number:
Molecular Weight:
191.01
Beilstein:
973353
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

301 °C (lit.)

mp

151-154 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1cc(Cl)ccc1Cl

InChI

1S/C7H4Cl2O2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,(H,10,11)

InChI key

QVTQYSFCFOGITD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine J Duckett et al.
Rapid communications in mass spectrometry : RCM, 16(4), 245-247 (2002-01-30)
The use of directly coupled high performance liquid chromatography/inductively coupled plasma mass spectroscopy (HPLC/ICPMS) employing chlorine ((35)Cl/(37)Cl) detection has been investigated with respect to the detection and quantitation of the drugs diclofenac and chlorpromazine. By integration of peak areas in
Heinz F Schöler et al.
Chemosphere, 60(11), 1505-1512 (2005-08-09)
The occurrence of halogenated organic compounds measured as a sum parameter and the evidence of chlorinated benzoic acids in four carbonaceous meteorites (Cold Bokkeveld, Murray, Murchison and Orgueil) from four independent fall events is reported. After AOX (Adsorbable organic halogen)
G D Griffith et al.
Applied and environmental microbiology, 58(1), 409-411 (1992-01-01)
Desulfomonile tiedjei DCB-1 is a strict anaerobe capable of reductively dechlorinating meta-chlorobenzoates. To probe the mechanism of this aryl dechlorination, we incubated cell suspensions of D. tiedjei in D2O and with 2,5-dichlorobenzoate. The deuterium was incorporated into the dechlorination product
Xiaopeng Xuan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1663-1668 (2011-06-15)
In this paper, experimental and theoretical studies on the molecular structure and vibrational spectra of methyl 2,5-dichlorobenzoate (MDCB) are presented. Fourier transform infrared and Raman spectra of the title molecule in the solid phase were recorded and analyzed. The geometrical
L Pavlû et al.
Journal of applied microbiology, 87(3), 381-386 (1999-10-30)
Two polychlorinated biphenyl-contaminated sites in the Czech Republic, a soil at Zamberk and a sediment sludge at Milevsko, were screened for the presence of chlorobenzoate degraders. Sixteen different chlorobenzoate degraders were isolated from the soil compared with only three strains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service