Skip to Content
Merck
All Photos(1)

Key Documents

24-4380

Sigma-Aldrich

Potassium citrate tribasic monohydrate

SAJ special grade, ≥99.0%

Synonym(s):

Citric acid tripotassium salt, Tripotassium citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOK)(CH2COOK)2 · H2O
CAS Number:
Molecular Weight:
324.41
Beilstein:
3924344
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

8.0-9.5

mp

275 °C (dec.) (lit.)

SMILES string

O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3K.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;1H2/q;3*+1;/p-3

InChI key

PJAHUDTUZRZBKM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeynep Yarbaşi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1304-1307 (2011-06-07)
An EPR and optical studies of VO2+ doped potassium dihydrogen citrate (PDHC) single crystals have been carried out at room temperature. It crystallizes in triclinic symmetry with the unit cell dimensions: a=11.343Å, b=13.078Å, c=6.272Å, α=89.79°, β=94.36°, γ=104.2°. The angular variation
Arun Gopalakrishnan et al.
Clinical and experimental nephrology, 15(5), 745-748 (2011-06-01)
Fanconi-Bickel syndrome is a rare autosomal recessive disorder characterized by hepatorenal glycogen accumulation, proximal renal tubular dysfunction and impaired utilization of glucose and galactose. Most cases have been reported from Europe, Japan, Turkey and the Mediterranean belt. We report a
Kendall F Moseley et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 28(3), 497-504 (2012-09-20)
The dietary acid load created by the typical Western diet may adversely impact the skeleton by disrupting calcium metabolism. Whether neutralizing dietary acid with alkaline potassium salts results in sustained improvements in calcium balance remains controversial. In this randomized, double-blind
Sigrid Jehle et al.
The Journal of clinical endocrinology and metabolism, 98(1), 207-217 (2012-11-20)
The acid load imposed by a modern diet may play an important role in the pathophysiology of osteoporosis. Our objective was to evaluate the skeletal efficacy and safety and the effect on fracture prediction of K-citrate to neutralize diet-induced acid
Asghar Rastegar
Clinical journal of the American Society of Nephrology : CJASN, 6(10), 2516-2521 (2011-09-17)
Hypokalemic paralysis represents a medical emergency requiring both rapid diagnosis and treatment. In this Attending Rounds a patient with hypokalemia and metabolic acidosis is presented to emphasize the role of routine laboratory studies in the assessment of such patients so

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service