Skip to Content
Merck
All Photos(1)

Key Documents

ST1606

Sigma-Aldrich

Anti-ABCG1 Mouse mAb (2E11)

liquid, clone 2E11, Calbiochem®

Synonym(s):

Anti-ATP-Binding Cassette G1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E11, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCG1(9619)

General description

Anti-ABCG1, mouse monoclonal antibody, clone 2E11, recognizes the ABCG1 protein. It is validated for use in ELISA.
Purified mouse monoclonal antibody. Recognizes the ABCG1 protein.
Recognizes the ABCG1 protein.

Immunogen

A recombinant polypeptide corresponding to amino acids 21-130 of human ABCG1, ecpressed as a GST fusion protein

Application

ELISA (see comments)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.19.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service