Skip to Content
Merck
All Photos(1)

Key Documents

HG00BX15

Millipore

Potassium tellurite

1% (solution)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K2TeO3
CAS Number:
Molecular Weight:
253.79
UNSPSC Code:
41171614

Pricing and availability is not currently available.

packaging

pkg of 10 ea (HG00BX15.PK)

concentration

1% (solution)

storage temp.

15-25°C

SMILES string

O=[Te](O)O

InChI

1S/H2O3Te/c1-4(2)3/h(H2,1,2,3)

InChI key

SITVSCPRJNYAGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service