Skip to Content
Merck
All Photos(1)

Key Documents

34856-M

Sigma-Aldrich

Dichloromethane

suitable for HPLC, ≥99.8%, contains amylene as stabilizer

Synonym(s):

methylene chloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.8%
technique(s):
HPLC: suitable
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

1223 °F

contains

amylene as stabilizer

expl. lim.

22 %

technique(s)

HPLC: suitable

impurities

free chlorine, in accordance
≤0.001% free acid (as HCl)
≤0.001% non-volatile matter
≤0.02% water (Karl Fischer)

color

APHA: ≤10

transmittance

235 nm, ≥20%
240 nm, ≥50%
245 nm, ≥80%
260 nm, ≥98%

refractive index

n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

absorption

in accordance

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane has been tested as a solvent medium for dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml. Role of the quantity of TiO2 loading on activated carbon support on dichloromethane photodecomposition has been investigated.

Application

Dichloromethane may be used in ultrasonic baths for cleaning and protection of aluminum coating over atactic-PMMA (poly(methyl methacrylate)) for dielectric studies.

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.
Important notice
This product will be phased out towards early 2019. There is a replacement product available : 34856

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service