Skip to Content
Merck
All Photos(1)

Key Documents

07-543

Sigma-Aldrich

Anti-p19 ARF Antibody

Upstate®, from rabbit

Synonym(s):

Anti-ARF, Anti-CDK4I, Anti-CDKN2, Anti-CMM2, Anti-INK4, Anti-INK4A, Anti-MLM, Anti-MTS-1, Anti-MTS1, Anti-P14, Anti-P14ARF, Anti-P16, Anti-P16-INK4A, Anti-P16INK4, Anti-P16INK4A, Anti-P19, Anti-P19ARF, Anti-TP16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

p19 ARF

Immunogen

Full length mouse p19 ARF fused to GST.

Application

Detect p19 ARF with Anti-p19 ARF Antibody (Rabbit Polyclonal Antibody), that has been shown to work in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Routinely evaluated by immunoblot with lysates from p53 knockout MEF cells.

Target description

19kDa

Physical form

ImmunoAffinity Purified

Storage and Stability

2 years at -20°C from date of shipment

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for mingels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shubhankar Suman et al.
Aging, 5(8), 607-622 (2013-08-10)
Despite recent epidemiological evidences linking radiation exposure and a number of human ailments including cancer, mechanistic understanding of how radiation inflicts long-term changes in cerebral cortex, which regulates important neuronal functions, remains obscure. The current study dissects molecular events relevant
p19ARF directly and differentially controls the functions of c-Myc independently of p53.
Ying Qi, Mark A Gregory, Zhaoliang Li, Jeffrey P Brousal, Kimberly West, Stephen R Hann
Nature null
Hui-Yi Kua et al.
Nature cell biology, 14(7), 727-737 (2012-06-26)
Defects in stem cell renewal or progenitor cell expansion underlie ageing-related diseases such as osteoporosis. Yet much remains unclear about the mechanisms regulating progenitor expansion. Here we show that the tyrosine kinase c-Abl plays an important role in osteoprogenitor expansion.
S Kitajima et al.
Oncogene, 30(6), 737-750 (2010-10-05)
The reversion-inducing cysteine-rich protein with Kazal motifs (RECK) gene had been isolated as an antagonist to RAS signaling; however, the mechanism of its action is not clear. In this study, the effect of loss of RECK function was assessed in
Nicolas Jullien et al.
PloS one, 10(3), e0120010-e0120010 (2015-03-31)
To test the role of wtPIT-1 (PITWT) or PIT-1 (R271W) (PIT271) in somatolactotroph cells, we established, using inducible lentiviral vectors, sublines of GH4C1 somatotroph cells that allow the blockade of the expression of endogenous PIT-1 and/or the expression of PITWT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service