Z743263
Julabo® bath tubing
tubing insulation, I.D. × L 18 mm × 1 m , for use with 12 mm I.D. reinforced tubing
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8930412
I.D. × L
18 mm × 1 m , for use with 12 mm I.D. reinforced tubing
Related Categories
General description
Flexible connection of the application to the temperature control instrument for optimal use in the respective temperature range.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)