Skip to Content
Merck
All Photos(2)

Key Documents

747459

Sigma-Aldrich

Iron oxide(II,III) magnetic nanopowder

10 nm avg. part. size (TEM), NHS ester functionalized

Synonym(s):

Fe NP NHS, FexOy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

form

nanopowder
powder

composition

Fe, ~1.25 % (w/w)

magnetization

>45 emu/g (at room temperature; under 4500 Oe)

color

brown

avg. part. size

10 nm (TEM)

functional group

NHS ester (functionalised)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Ocean Nanotech; LLC.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Characterizing the Structure and Defect Concentration of ZnO Nanoparticles in a Colloidal Solution.
Rossi T, et al.
The Journal of Physical Chemistry C, 118(33), 19422-19430 (2014)
The preparation of magnetic nanoparticles for applications in biomedicine.
Tartaj P, et al.
Journal of Physics D: Applied Physics, 36(13), R182-R182 (2003)

Articles

Professor Yadong Yin (University of California Riverside, USA) examines both direct (thermal decomposition, solvothermal, hydrothermal) and indirect (templated) synthesis methods of magnetite nanocrystals and reviews in detail the landscape of these various synthetic methods for magnetite nanocrystal and their applications in magnetic assembly, magnetic hyperthermia, and Li-Ion batteries.

The recent emergence of a number of highly functional nanomaterials has enabled new approaches to the understanding, diagnosis, and treatment of cancer.

Biomaterials science involves the design and fabrication of smart materials for studying, directing, or mimicking biology. For successful integration of biomaterials in biological research, a meaningful understanding of biological systems is required.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service