Skip to Content
Merck
All Photos(1)

Key Documents

733709

Sigma-Aldrich

Diisobutylaluminum deuteride solution

0.7 M in toluene, 98 atom % D

Synonym(s):

DIBAL-D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]2AlD
Molecular Weight:
143.22
MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

form

liquid

concentration

0.7 M in toluene

density

0.851 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H][Al](CC(C)C)CC(C)C

InChI

1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;/i;;;1+1

InChI key

AZWXAPCAJCYGIA-SGNQUONSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisobutylaluminum deuteride solution is isotopically labeled reagent of DIBAL-hydride.

Application

DIBAL-D is used in identification of stereochemistry of alkene products from the reduction process of acetylene derivatives.It is also used in the synthesis of 1-deuterioaldehydes from esters.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service