HPLC Analysis of Luciferin Enantiomers on CHIRALPAK® AGP
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
CHIRALPAK AGP, 10 cm x 4 mm I.D., 5 μm particles (58150AST)
mobile phase
10 mM sodium phosphate, pH 6.0
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 225 nm
sample
luciferin
Description
Legal Information
CHIRALPAK is a registered trademark of Daicel Corp.