GC Analysis of Barbiturate Enantiomers on β-DEX™ 120
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
β-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24304)
oven
210 °C
inj. temp.
300 °C
detector
FID, 300 °C
carrier gas
helium, 20 cm/sec @ 275 °C
injection
100:1 split
Description
Legal Information
DEX is a trademark of Sigma-Aldrich Co. LLC