GC Analysis of Water (100% Standard) on Watercol™ 1460
CONDITIONS
column
Watercol 1460, 30 m x 0.25 mm I.D., 0.20 μm (29695-U)
oven
60 °C (12 min)
inj. temp.
250 °C
detector
TCD, 200 °C
carrier gas
helium, 26 cm/sec
injection
0.1 μL, 100:1 split
liner
4 mm I.D., split type, cup design
sample
water
Description
Analysis Note
The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. This application shows a calibration standard (100% water) on Watercol 1460.
Legal Information
Watercol is a trademark of Sigma-Aldrich Co. LLC